3-糠醛 ≥98.0% (通过GC)
供应商: TCI EUROPE
|
Danger
|
Synonyms:
3-呋喃甲醛
TCIAF0354-10GEA
0
CNY
TCIAF0354-10G
3-糠醛 ≥98.0% (通过GC)
3-糠醛
分子式:
C₅H₄O₂ 分子量: 96,09 g/mol 沸点: 144°C 闪点: 48°C 存储条件: Refrigerator |
MDL编号:
MFCD00010424 CAS号: 498-60-2 UN号: 1199 ADR: 6.1,II |
产品技术规格
Appearance | Colorless to Brown clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...