5-(4-Fluorophenoxy)valeric acid ≥98.0% (通过GC,滴定分析)

供应商: TCI EUROPE
Warning

TCIAF0410-5GEA 0 CNY
TCIAF0410-5G
5-(4-Fluorophenoxy)valeric acid ≥98.0% (通过GC,滴定分析)
5-(4-Fluorophenoxy)valeric acid
分子式: C₁₁H₁₃FO₃
分子量: 212,22 g/mol
MDL编号: MFCD02093469
CAS号: 347867-75-8

现在订购

产品技术规格

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 79.0 to 82.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR