4-(4-Fluorophenylethynyl)phenol ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAF0423-1GEA 760 CNY
TCIAF0423-1G
4-(4-Fluorophenylethynyl)phenol ≥98.0% (通过GC)
4-(4-Fluorophenylethynyl)phenol
分子式: C₁₄H₉FO
分子量: 212,22 g/mol
MDL编号: MFCD02093472
CAS号: 197770-48-2

现在订购

产品技术规格

Appearance White to Light red to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 147.0 to 151.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR