4-Formylphenyl-β-D-allopyranoside ≥98.0% (通过HPLC)
供应商: TCI EUROPE
TCIAF0542-5GEA
0
CNY
TCIAF0542-5G
4-Formylphenyl-β-D-allopyranoside ≥98.0% (通过HPLC)
4-Formylphenyl-beta-D-allopyranoside
分子式:
C₁₃H₁₆O₇ 分子量: 284,26 g/mol 溶点: 190°C |
CAS号:
80154-34-3 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Specific rotation [a]20/D | -95.0 to -115.0 deg (C=0.8, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...