3-Fluoro-4-methylpyridine ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAF0852-1GEA 275 CNY
TCIAF0852-1G
3-Fluoro-4-methylpyridine ≥98.0% (通过GC)
3-Fluoro-4-methylpyridine
分子式: C₆H₆FN
分子量: 111,12 g/mol
沸点: 136°C
MDL编号: MFCD04114226
CAS号: 399-88-2
UN号: 1993
ADR: 3,III

现在订购

产品技术规格

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR