2,4-Heptadien-6-one ≥95.0% (通过GC) stabilised

供应商: TCI EUROPE
Warning


Stabilization: Stabilised with hydroquinone (HQ)

TCIAH0022-5GEA 2000 CNY
TCIAH0022-5G
2,4-Heptadien-6-one ≥95.0% (通过GC) stabilised
2,4-Heptadien-6-one
分子式: C₇H₁₀O
分子量: 110,16 g/mol
沸点: 88°C
MDL编号: MFCD00059238
CAS号: 3916-64-1

现在订购

产品技术规格

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 95.0 % (total of isomer)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR