2,4,6-Tribromophenyl hexanoate ≥97.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAH0113-25GEA 0 CNY
TCIAH0113-25G
2,4,6-Tribromophenyl hexanoate ≥97.0% (通过GC)
2,4,6-Tribromophenyl hexanoate
分子式: C₁₂H₁₃Br₃O₂
分子量: 428,95 g/mol
MDL编号: MFCD00059464
CAS号: 16732-09-5

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR