1-(2-Hydroxyethyl)-4-(3-hydroxypropyl)piperidine ≥98.0% (通过GC,滴定分析)

供应商: TCI EUROPE
Warning

TCIAH0360-25GEA 475 CNY
TCIAH0360-25G
1-(2-Hydroxyethyl)-4-(3-hydroxypropyl)piperidine ≥98.0% (通过GC,滴定分析)
1-(2-Hydroxyethyl)-4-(3-hydroxypropyl)piperidine
分子式: C₁₀H₂₁NO₂
分子量: 187,28 g/mol
MDL编号: MFCD00047481
CAS号: 19780-85-9
EINECS: 243-305-6

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 46.0 to 51.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR