2-Undecylimidazoline ≥90.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAH0382-25GEA 0 CNY
TCIAH0382-25G
2-Undecylimidazoline ≥90.0% (通过GC)
2-Undecylimidazoline
分子式: C₁₄H₂₈N₂
MDL编号: MFCD00070626
CAS号: 10443-61-5

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 90.0 %
Melting point 79.0 to 83.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR