(2-Hydroxyethyl)triethylammonium Iodide ≥98.0% (通过滴定分析)

供应商: TCI EUROPE
Warning

TCIAH0407-10GEA 0 CNY
TCIAH0407-10G
(2-Hydroxyethyl)triethylammonium Iodide ≥98.0% (通过滴定分析)
(2-Hydroxyethyl)triethylammonium Iodide
分子式: C₈H₂₀INO
分子量: 273,16 g/mol
MDL编号: MFCD00031691
CAS号: 5957-17-5

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR