4'-Hexyloxybenzylidene-4-cyanoaniline ≥98.0% (通过滴定分析)

供应商: TCI EUROPE
Warning

TCIAH0419-1GEA 340 CNY
TCIAH0419-1G
4'-Hexyloxybenzylidene-4-cyanoaniline ≥98.0% (通过滴定分析)
4'-Hexyloxybenzylidene-4-cyanoaniline
分子式: C₂₀H₂₂N₂O
MDL编号: MFCD00059589
CAS号: 35280-78-5
EINECS: 252-486-0

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR