3-Hydroxyphenylurea ≥97.0% (以总氮计)

供应商: TCI EUROPE
Warning

TCIAH0438-25GEA 0 CNY
TCIAH0438-25G
3-Hydroxyphenylurea ≥97.0% (以总氮计)
3-Hydroxyphenylurea
分子式: C₇H₈N₂O₂
分子量: 152,15 g/mol
MDL编号: MFCD00007945
CAS号: 701-82-6
EINECS: 211-860-3

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (with Total Nitrogen) min. 97.0 %
Melting point 177.0 to 181.0 deg-C
Solubility in hot Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR