5-Heptyn-3-ol ≥97.0% (通过GC)

供应商: TCI EUROPE

TCIAH0459-5MLEA 0 CNY
TCIAH0459-5ML
5-Heptyn-3-ol ≥97.0% (通过GC)
5-Heptyn-3-ol
分子式: C₇H₁₂O
分子量: 112,17 g/mol
MDL编号: MFCD00046680
CAS号: 19781-82-9

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR