3-Hydroxyanthranilic acid hydrochloride ≥98.0% (通过HPLC,滴定分析)

供应商: TCI EUROPE

TCIAH0495-1GEA 0 CNY
TCIAH0495-1G
3-Hydroxyanthranilic acid hydrochloride ≥98.0% (通过HPLC,滴定分析)
3-Hydroxyanthranilic acid hydrochloride
分子式: C₇H₇NO₃
分子量: 189,60 g/mol
MDL编号: MFCD00012965
CAS号: 4920-81-4
EINECS: 225-550-0

现在订购

产品技术规格

Appearance White to Light red to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR