2-Ethyl-3-hydroxy-4-pyrone

供应商: TCI EUROPE
Warning

Synonyms: 乙基麦芽酚2-乙基-3-羟基-4H-吡喃-4-酮

TCIAH0550-25GEA 0 CNY
TCIAH0550-25G
2-Ethyl-3-hydroxy-4-pyrone
乙基麦芽酚
分子式: C₇H₈O₃
分子量: 140,14 g/mol
MDL编号: MFCD00059795
CAS号: 4940-11-8
EINECS: 225-582-5

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Neutralization titration) min. 99.0 %
Melting point 89.0 to 93.0 deg-C
Solubility in hot Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR