2-Heptynoic acid ≥97.0% (通过滴定分析)
供应商: TCI EUROPE
|
Danger
|
TCIAH0664-1MLEA
0
CNY
TCIAH0664-1ML
2-Heptynoic acid ≥97.0% (通过滴定分析)
2-Heptynoic acid
分子式:
C₇H₁₀O₂ 分子量: 126,16 g/mol 沸点: 141°C |
MDL编号:
MFCD00014372 CAS号: 1483-67-6 EINECS: 216-048-2 UN号: 3265 ADR: 8,III |
产品技术规格
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...