(2-羟乙基)脲 ≥98.0% (以总氮计)

供应商: TCI EUROPE

TCIAH0700-25GEA 0 CNY
TCIAH0700-25G
(2-羟乙基)脲 ≥98.0% (以总氮计)
(2-羟乙基)脲
分子式: C₃H₈N₂O₂
分子量: 104,11 g/mol
MDL编号: MFCD00059080
CAS号: 2078-71-9

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 91.0 to 95.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR