1-(3-Hydroxypropyl)pyrrole ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAH0827-1GEA 0 CNY
TCIAH0827-1G
1-(3-Hydroxypropyl)pyrrole ≥98.0% (通过GC)
1-(3-Hydroxypropyl)pyrrole
分子式: C₇H₁₁NO
分子量: 125,17 g/mol
沸点: 231°C
存储条件: Refrigerator
MDL编号: MFCD00191453
CAS号: 50966-69-3

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR