4-Hydroxy-1-(2-hydroxyethyl)-2,2,6,6-tetramethylpiperidine ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAH0931-5GEA 0 CNY
TCIAH0931-5G
4-Hydroxy-1-(2-hydroxyethyl)-2,2,6,6-tetramethylpiperidine ≥98.0% (通过GC)
4-Hydroxy-1-(2-hydroxyethyl)-2,2,6,6-tetramethylpiperidine
分子式: C₁₁H₂₃NO₂
分子量: 201,31 g/mol
MDL编号: MFCD01321174
CAS号: 52722-86-8
EINECS: 258-132-1

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 184.0 to 187.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR