2-羟基-4-邻苯二甲酰亚氨基丁酸 ≥98.0% (通过HPLC,滴定分析)

供应商: TCI EUROPE
Warning

TCIAH0933-5GEA 0 CNY
TCIAH0933-5G
2-羟基-4-邻苯二甲酰亚氨基丁酸 ≥98.0% (通过HPLC,滴定分析)
2-羟基-4-邻苯二甲酰亚氨基丁酸
分子式: C₁₂H₁₁NO₅
分子量: 249,22 g/mol
MDL编号: MFCD00192213
CAS号: 48172-10-7

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 150.0 to 154.0 deg-C
Specific rotation [a]20/D +5.0 to +7.0 deg (C=1, Methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR