(Hydrazinocarbonyl)ferrocene ≥95.0% (通过HPLC,滴定分析) for HPLC labeling
供应商: TCI EUROPE
|
Warning
|
TCIAH0941-1GEA
0
CNY
TCIAH0941-1G
(Hydrazinocarbonyl)ferrocene ≥95.0% (通过HPLC,滴定分析) for HPLC labeling
(Hydrazinocarbonyl)ferrocene
分子式:
C₁₁H₁₂FeN₂O 分子量: 244,08 g/mol |
MDL编号:
MFCD01321175 CAS号: 12153-28-5 |
产品技术规格
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Nonaqueous Titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...