4-Methylpyrimidin-2-ol hydrochloride ≥98.0% (通过滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAH0943-25GEA
0
CNY
TCIAH0943-25G
4-Methylpyrimidin-2-ol hydrochloride ≥98.0% (通过滴定分析)
4-Methylpyrimidin-2-ol hydrochloride
分子式:
C₅H₇ClN₂O 分子量: 146,58 g/mol |
MDL编号:
MFCD00151347 CAS号: 5348-51-6 EINECS: 226-306-6 |
产品技术规格
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...