4-(羟基甲基)苯氧基乙酸 ≥98.0% (通过滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAH0961-1GEA
0
CNY
TCIAH0961-1G
4-(羟基甲基)苯氧基乙酸 ≥98.0% (通过滴定分析)
4-(羟基甲基)苯氧基乙酸
分子式:
C₉H₁₀O₄ 分子量: 182,2 g/mol |
CAS号:
68858-21-9 |
产品技术规格
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 109.0 to 112.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...