3-(4-Hydroxy-3-methoxyphenyl)-1-propanol ≥98.0% (通过HPLC)
供应商: TCI EUROPE
|
Warning
|
TCIAH1011-1GEA
0
CNY
TCIAH1011-1G
3-(4-Hydroxy-3-methoxyphenyl)-1-propanol ≥98.0% (通过HPLC)
3-(4-Hydroxy-3-methoxyphenyl)-1-propanol
分子式:
C₁₀H₁₄O₃ 分子量: 182,22 g/mol |
MDL编号:
MFCD00016571 CAS号: 2305-13-7 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 59.0 to 65.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...