2-Hydroxyisobutyramide ≥98.0% (以总氮计)

供应商: TCI EUROPE
Warning

TCIAH1181-25GEA 840 CNY
TCIAH1181-25G
2-Hydroxyisobutyramide ≥98.0% (以总氮计)
2-Hydroxyisobutyramide
分子式: C₄H₉NO₂
沸点: 260°C
MDL编号: MFCD08063365
CAS号: 13027-88-8
EINECS: 235-890-1

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 91.0 to 96.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR