1,1,1,3,5,5,5-Heptamethyl-3-(3-glycidyloxypropyl)trisiloxane ≥96.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
TCIAH1266-5GEA
0
CNY
TCIAH1266-5G
1,1,1,3,5,5,5-Heptamethyl-3-(3-glycidyloxypropyl)trisiloxane ≥96.0% (通过GC)
1,1,1,3,5,5,5-Heptamethyl-3-(3-glycidyloxypropyl)trisiloxane
分子式:
C₁₃H₃₂O₄Si₃ 分子量: 336,65 g/mol 沸点: 98°C |
MDL编号:
MFCD00053926 CAS号: 7422-52-8 EINECS: 231-045-6 |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...