外-6-羟基托品酮 ≥98.0% (通过GC,滴定分析)

供应商: TCI EUROPE

TCIAH1289-1GEA 0 CNY
TCIAH1289-1G
外-6-羟基托品酮 ≥98.0% (通过GC,滴定分析)
外-6-羟基托品酮
分子式: C₈H₁₃NO₂
分子量: 155,20 g/mol
MDL编号: MFCD00135648
CAS号: 5932-53-6
EINECS: 227-677-7

现在订购

产品技术规格

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 121.0 to 125.0 deg-C
Specific rotation -1.0 to +1.0 deg (C=1, water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR