1-Hydroxytetraphenylcylclopentadienyl(tetraphenyl-2,4-cyclopentadien-1-one)-mu-hydrotetracarbonyldiruthenium(II)
供应商: TCI EUROPE
Synonyms:
四羰基-μ-氢[(1,2,3,4,5-H)-1-羟基-2,3,4,5-四苯基-2,4-环戊二烯-1-yl][(1,2,3,4,5-H)-1-羟基-2,3,4,5-四苯基-2,4-环戊二烯-1-基]二钌
, 2,4-环戊二烯-1-酮 2,3,4,5-四苯基钌络合物
, Shvo 催化剂
TCIAH1322-100MGEA
0
CNY
TCIAH1322-100MG
1-Hydroxytetraphenylcylclopentadienyl(tetraphenyl-2,4-cyclopentadien-1-one)-mu-hydrotetracarbonyldiruthenium(II)
1-羟基四苯基-环戊二烯基(四苯基-2,4-环戊二烯-1-酮)-μ-氢四羰基二钌(II)
分子式:
C₆₂H₄₂O₆Ru₂ 分子量: 1085,15 g/mol |
MDL编号:
MFCD03840556 CAS号: 104439-77-2 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Elemental analysis (Carbon) | 67.0 to 71.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...