4'-Iodo-4-dimethylaminoazobenzene ≥97.0% (通过滴定分析)

供应商: TCI EUROPE
Warning

TCIAI0057-5GEA 0 CNY
TCIAI0057-5G
4'-Iodo-4-dimethylaminoazobenzene ≥97.0% (通过滴定分析)
4'-Iodo-4-dimethylaminoazobenzene
分子式: C₁₄H₁₄IN₃
分子量: 351,19 g/mol
MDL编号: MFCD00059492
CAS号: 3805-67-2

现在订购

产品技术规格

Appearance Light yellow to Brown powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 162.0 to 166.0 deg-C
Solubility in Toluene ithin almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR