4-Isobutylmorpholine ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAI0254-25MLEA 0 CNY
TCIAI0254-25ML
4-Isobutylmorpholine ≥98.0% (通过GC)
4-Isobutylmorpholine
分子式: C₈H₁₇NO
分子量: 143,23 g/mol
沸点: 167°C
闪点: 47°C
MDL编号: MFCD00023382
CAS号: 10315-98-7
EINECS: 233-696-1
UN号: 1993
ADR: 3,III

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR