2-(Isobutylthio)ethanol ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAI0264-5MLEA 930 CNY
TCIAI0264-5ML
2-(Isobutylthio)ethanol ≥98.0% (通过GC)
2-(Isobutylthio)ethanol
分子式: C₆H₁₄OS
分子量: 134,24 g/mol
沸点: 88°C
MDL编号: MFCD00014040
CAS号: 42779-10-2

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR