Isatin-3-oxime ≥98.0% (通过HPLC,滴定分析) for metal analysis

供应商: TCI EUROPE

TCIAI0273-5GEA 0 CNY
TCIAI0273-5G
Isatin-3-oxime ≥98.0% (通过HPLC,滴定分析) for metal analysis
Isatin-3-oxime
分子式: C₈H₆N₂O₂
分子量: 162,15 g/mol
MDL编号: MFCD00014568
CAS号: 607-28-3
EINECS: 210-132-2

现在订购

产品技术规格

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Sensitiveness to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR