4-Isopropoxy-2-butanone ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAI0283-5MLEA 0 CNY
TCIAI0283-5ML
4-Isopropoxy-2-butanone ≥98.0% (通过GC)
4-Isopropoxy-2-butanone
分子式: C₇H₁₄O₂
分子量: 130,19 g/mol
沸点: 75°C
MDL编号: MFCD00059358
CAS号: 32541-58-5
UN号: 1993
ADR: 3,III

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR