4-Isobutoxy-2-butanone ≥95.0% (通过GC)

供应商: TCI EUROPE

TCIAI0286-5MLEA 0 CNY
TCIAI0286-5ML
4-Isobutoxy-2-butanone ≥95.0% (通过GC)
4-Isobutoxy-2-butanone
分子式: C₈H₁₆O₂
分子量: 144,21 g/mol
沸点: 181°C
MDL编号: MFCD00059348
CAS号: 31576-33-7

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR