1-Isopropyl-3-pyrrolidinol ≥96.0% (通过GC,滴定分析)

供应商: TCI EUROPE

TCIAI0430-5GEA 0 CNY
TCIAI0430-5G
1-Isopropyl-3-pyrrolidinol ≥96.0% (通过GC,滴定分析)
1-Isopropyl-3-pyrrolidinol
分子式: C₇H₁₅NO
分子量: 129,20 g/mol
沸点: 118°C
MDL编号: MFCD00191525
CAS号: 42729-56-6

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR