3-Buten-1-ylIsothiocyanate ≥96.0% (通过GC)

供应商: TCI EUROPE
Danger

TCIAI0443-1GEA 0 CNY
TCIAI0443-1G
3-Buten-1-ylIsothiocyanate ≥96.0% (通过GC)
3-Buten-1-ylIsothiocyanate
分子式: C₅H₇NS
分子量: 113,18 g/mol
沸点: 77°C
MDL编号: MFCD00191660
CAS号: 3386-97-8
EINECS: 222-209-8
UN号: 1992
ADR: 3,III

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR