3,4-O-异亚丙基-D-甘露糖醇 ≥96.0% (通过GC)

供应商: TCI EUROPE

TCIAI0489-1GEA 0 CNY
TCIAI0489-1G
3,4-O-异亚丙基-D-甘露糖醇 ≥96.0% (通过GC)
3,4-O-异亚丙基-D-甘露糖醇
分子式: C₉H₁₈O₆
分子量: 222,24 g/mol
MDL编号: MFCD00075122
CAS号: 3969-84-4

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Melting point 83.0 to 88.0 deg-C
Specific rotation [a]20/D +27.0 to +31.0 deg (C=3, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR