L-(-)-手性肌醇 ≥98.0% (通过HPLC)

供应商: TCI EUROPE

TCIAI0633-200MGEA 0 CNY
TCIAI0633-200MG
L-(-)-手性肌醇 ≥98.0% (通过HPLC)
L-(-)-手性肌醇
分子式: C₆H₁₂O₆
分子量: 180,16 g/mol
溶点: 240°C
存储条件: Freezer
MDL编号: MFCD00799556
CAS号: 551-72-4
EINECS: 209-000-7

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D -60.0 to -70.0 deg (C=1, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR