2',3'-O-Isopropylideneinosine ≥98.0% (通过HPLC)

供应商: TCI EUROPE

TCIAI0704-5GEA 0 CNY
TCIAI0704-5G
2',3'-O-Isopropylideneinosine ≥98.0% (通过HPLC)
2',3'-O-Isopropylideneinosine
分子式: C₁₃H₁₆N₄O₅
分子量: 308,29 g/mol
MDL编号: MFCD00038002
CAS号: 2140-11-6
EINECS: 218-388-7

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 96.0 %
Specific rotation [a]20/D -61.0 to -71.0 deg (C=0.8, Methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR