5-(Isopropylamino)pentanol ≥98.0% (通过滴定分析)

供应商: TCI EUROPE
Warning

TCIAI0757-1GEA 410 CNY
TCIAI0757-1G
5-(Isopropylamino)pentanol ≥98.0% (通过滴定分析)
5-(Isopropylamino)pentanol
分子式: C₈H₁₉NO
分子量: 145,25 g/mol
沸点: 125°C
MDL编号: MFCD14708174
CAS号: 40447-21-0

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 42.0 to 46.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR