4-Iodo-4',4''-dimethoxytriphenylamine ≥98.0% (通过HPLC,总氮)
供应商: TCI EUROPE
|
Warning
|
TCIAI0776-1GEA
1475
CNY
TCIAI0776-1G
4-Iodo-4',4''-dimethoxytriphenylamine ≥98.0% (通过HPLC,总氮)
4-Iodo-4',4''-dimethoxytriphenylamine
分子式:
C₂₀H₁₈INO₂ 分子量: 431,27 g/mol |
CAS号:
201802-15-5 |
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 109.0 to 113.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...