6-Iodo-4-hydroxyquinazoline ≥97.0% (通过GC,滴定分析)

供应商: TCI EUROPE
Warning

TCIAI0832-5GEA 0 CNY
TCIAI0832-5G
6-Iodo-4-hydroxyquinazoline ≥97.0% (通过GC,滴定分析)
6-Iodo-4-hydroxyquinazoline
分子式: C₈H₅IN₂O
分子量: 272,05 g/mol
溶点: 271°C
MDL编号: MFCD00460007
CAS号: 16064-08-7

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR