4-甲基-2-氧代戊酸 ≥95.0% (通过GC,滴定分析)
供应商: TCI EUROPE
|
Danger
|
Synonyms:
酮亮氨酸
, 4-甲基-2-氧基戊酸
, α-酮异己酸
, 2-羰基异己酸
TCIAK0025-5GEA
0
CNY
TCIAK0025-5G
4-甲基-2-氧代戊酸 ≥95.0% (通过GC,滴定分析)
4-甲基-2-氧代戊酸
分子式:
C₆H₁₀O₃ 分子量: 130,14 g/mol 沸点: 85°C |
MDL编号:
MFCD00066204 CAS号: 816-66-0 EINECS: 212-435-5 UN号: 3265 ADR: 8,III |
产品技术规格
Appearance | White to Yellow to Green clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...