Calcium lactobionate hydrate ≥97.0% (通过滴定分析)

供应商: TCI EUROPE

TCIAL0006-25GEA 0 CNY
TCIAL0006-25G
Calcium lactobionate hydrate ≥97.0% (通过滴定分析)
Calcium lactobionate hydrate
分子式: C₁₂H₂₂O₁₂
分子量: 754,65 g/mol
MDL编号: MFCD00135895
CAS号: 5001-51-4
EINECS: 225-668-2

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 97.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +22.0 to +25.0 deg (C=6, H2O)
Water 2.0 to 10.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR