4-(Methylamino)azobenzene

供应商: TCI EUROPE
Warning

TCIAM0143-5GEA 0 CNY
TCIAM0143-5G
4-(Methylamino)azobenzene
4-(Methylamino)azobenzene
分子式: C₁₃H₁₃N₃
MDL编号: MFCD00059198
CAS号: 621-90-9

现在订购

产品技术规格

Appearance Light yellow to Brown powder to crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR