2-Mercapto-5-thiazolidone ≥98.0% (通过HPLC,滴定分析)

供应商: TCI EUROPE

TCIAM0263-1GEA 995 CNY
TCIAM0263-1G
2-Mercapto-5-thiazolidone ≥98.0% (通过HPLC,滴定分析)
2-Mercapto-5-thiazolidone
分子式: C₃H₃NOS₂
分子量: 133,18 g/mol
存储条件: Freezer
MDL编号: MFCD00059745
CAS号: 6913-23-1

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR