2-Methyl-2-hexene ≥97.0% (通过GC)

供应商: TCI EUROPE
Danger

TCIAM0526-1GEA 630 CNY
TCIAM0526-1G
2-Methyl-2-hexene ≥97.0% (通过GC)
2-Methyl-2-hexene
分子式: C₇H₁₄
分子量: 98,19 g/mol
沸点: 95°C
闪点: -2°C
MDL编号: MFCD00027232
CAS号: 2738-19-4
UN号: 3295
ADR: 3,II

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR