2-[(4-Methoxybenzylidene)amino]fluorene ≥98.0% (通过滴定分析)

供应商: TCI EUROPE

TCIAM0586-5GEA 0 CNY
TCIAM0586-5G
2-[(4-Methoxybenzylidene)amino]fluorene ≥98.0% (通过滴定分析)
2-[(4-Methoxybenzylidene)amino]fluorene
分子式: C₂₁H₁₇NO
分子量: 299,37 g/mol
MDL编号: MFCD00019064
CAS号: 5424-78-2

现在订购

产品技术规格

Appearance Light yellow to Yellow to Orange powder to crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR