3,4-(亚甲二氧)肉桂酸,反式为主 ≥98.0% (通过HPLC,滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAM0634-25GEA
0
CNY
TCIAM0634-25G
3,4-(亚甲二氧)肉桂酸,反式为主 ≥98.0% (通过HPLC,滴定分析)
3,4-(亚甲二氧)肉桂酸,反式为主
分子式:
C₁₀H₈O₄ 分子量: 192,17 g/mol |
MDL编号:
MFCD00005837 CAS号: 2373-80-0 EINECS: 219-151-0 |
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Pyridine | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...