4-甲基-2-苯基咪唑 ≥93.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAM0669-25GEA 330 CNY
TCIAM0669-25G
4-甲基-2-苯基咪唑 ≥93.0% (通过GC)
4-甲基-2-苯基咪唑
分子式: C₁₀H₁₀N₂
分子量: 158,20 g/mol
MDL编号: MFCD00047015
CAS号: 827-43-0
EINECS: 212-571-5

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 93.0 %
Melting point 180.0 to 184.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR